Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Amersham™ Hybond™ -N+ Membranes Available on GSA/VA Contract for Federal Government customers only.

Use for the highest sensitivity in DNA and RNA blotting. GE Healthcare Life Sciences™ Amersham™ Hybond™ -N+ Membrane is a positively charged nylon membranes for use with radioactive and nonradioactive chemiluminescence and chemifluorescence detection.

Manufacturer:  GE Healthcare RPN82B

 View more versions of this product

Catalog No. 45-000-949


Add to cart

Description

Description

  • Pore size of 0.45µm with retention of nucleic acids >50bp
  • Binding capacity for nucleic acids 600µg/cm2
  • Suitable for high-density arrays
  • Excellent sensitivity in conventional Southern blotting
  • Nucleic acid samples may be fixed by UV or baking
  • Not suitable for alkali blotting and fixation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

50 Discs
Nylon, Positively Charged
Up to 600μg/cm2
Not Applicable
Disc
Southern blots; UV fixation
82 mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit