Learn More
GE Healthcare Amersham™ Hybond™-N Membranes Available on GSA/VA Contract for Federal Government customers only.
Use for high sensitivity applications in DNA and RNA blotting. GE Healthcare Life Sciences™Amersham™ Hybond™-N Membranes is a strong supported membrane that is inherently hydrophilic and requires no pre-wetting.
Manufacturer:  GE Healthcare RPN82N
 View more versions of this product
Catalog No. 45-000-953
Add to cart
Description
Description
  • Binding capacity for nucleic acids up to 600µm/cm2
  • Inherently hydrophilic and requires no prewetting
  • Nucleic acids can be crosslinked using UV light
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Disc
8.2cm
Up to 600μg/cm2
Nylon, Neutral
Southern and Northern blots; plaque lifts; UV fixation
82 mm