Learn More
GE Healthcare Amersham™ Hybond™ -NX Membranes
Neutral nylon membrane developed specifically for use with low hybridization buffer volumes and high-throughput applications
Manufacturer:  GE Healthcare RPN303T
 View more versions of this product
Catalog No. 45-000-933
Add to cart
Description
Description
  • Binding capacity for nucleic acids 600µg/cm2
  • High tensile strength withstands multiple reprobing
Specifications
Specifications
Roll
30 cm
Radioactive probes
Nylon, Highly Positively Charged
3 m
Up to 600μg/cm2