GE Healthcare Amersham™ Hybond™ -NX Membranes Available on GSA/VA Contract for Federal Government customers only.

Neutral nylon membrane developed specifically for use with low hybridization buffer volumes and high-throughput applications

Manufacturer: GE Healthcare RPN203T

 View more versions of this product

Catalog No. 45-000-855

  Check Availability
Add to Cart

    Specifications

    Material Nylon, Highly Positively Charged
    Binding Capacity Up to 600μg/cm2
    Format Roll
    Length (Metric) 20cm
    Width (Metric) 3m
    For Use With (Equipment) Radioactive probes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit