GE Healthcare Amersham™ Hybond™ PVDF Membranes: Rolls Available on GSA/VA Contract for Federal Government customers only.

Ideal for Western blotting applications where stripping and reprobing are necessary.  GE Healthcare Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls  are hydrophobic PVDF membranes.

Manufacturer: GE Healthcare 10600030

 View more versions of this product

Catalog No. 45-004-022

  Check Availability
Add to Cart

    Specifications

    Format Roll
    Grade SEQ 0.2μm
    For Use With (Application) Specifically designed for protein sequencing applications and solid phase amino acid analysis

    p>Supplied in rolls.

    • Hydrophobic PVDF membranes
    • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

    Amersham Hybond 0.2µm

    • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2µm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2µm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit