Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Amersham™ Hybond™ PVDF Membranes: Sheets

Ideal for Western blotting applications where stripping and reprobing are necessary

Manufacturer:  GE Healthcare 10600040

 View more versions of this product

Catalog No. 45-004-096

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Individual sheets.

  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2µm

  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2µm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2µm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

    Specifications

    Specifications

    5/Pk.
    PVDF
    26.5 x 60 cm
    Sheets (LFP)
    Ideal for Western blotting applications where stripping and reprobing are necessary
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit