GE Healthcare Amersham™ Hybond™ PVDF Membranes: Sheets Available on GSA/VA Contract for Federal Government customers only.

Ideal for Western blotting applications where stripping and reprobing are necessary

Manufacturer: GE Healthcare 10600092

 View more versions of this product

Catalog No. 45-004-071

  Check Availability
Add to Cart

    Specifications

    Material PVDF
    Format Sheets (Seq)
    Dimensions (L x W) 10 × 10cm
    For Use With (Application) Ideal for Western blotting applications where stripping and reprobing are necessary
    Quantity 10/Pk.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit