GE Healthcare Amersham™ Hybond™ -XL Membranes Available on GSA/VA Contract for Federal Government customers only.

Use for optimal signal-to-noise ratios when using radioactively labeled probes. GE Healthcare Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.

Manufacturer: GE Healthcare RPN303S

 View more versions of this product

Catalog No. 45-001-148

  Check Availability
Add to Cart

    Specifications

    Material Nylon, Highly Positively Charged
    Binding Capacity Up to 600μg/cm2
    Format Roll
    Length (Metric) 30cm
    Width (Metric) 3m
    For Use With (Equipment) Radioactive probes
    Quantity 1 Roll

    • Greater concentration and more even distribution of positive charge than other charged nylon membranes
    • Binding capacity for nucleic acids up to 600µg/cm2
    • Efficient binding of small fragments
    • Effective probe removal
    • Suitable for reprobing
    • Discs are also available

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit