GE Healthcare Amersham™ Hybond™ -XL Membranes Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: GE Healthcare RPN2222S

Catalog No. 45-001-152

  Check Availability
Add to Cart

    Specifications

    Material Nylon, Highly Positively Charged
    Binding Capacity Up to 600μg/cm2
    Format Sheet
    Length (Metric) 22.2cm
    Width (Metric) 22.2cm
    For Use With (Equipment) Radioactive probes
    Quantity 10 Sheets

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit