Learn More
GE Healthcare Amersham™ Hybond™ -XL Membranes
Use for optimal signal-to-noise ratios when using radioactively labeled probes. GE Healthcare Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.
Manufacturer:  GE Healthcare RPN2020S
 View more versions of this product
Catalog No. 45-001-147
Add to cart
Description
Description
  • Greater concentration and more even distribution of positive charge than other charged nylon membranes
  • Binding capacity for nucleic acids up to 600µg/cm2
  • Efficient binding of small fragments
  • Effective probe removal
  • Suitable for reprobing
  • Discs are also available
Specifications
Specifications
10 Sheets
Nylon, Highly Positively Charged
20 cm
Up to 600μg/cm2
Sheet
20 cm
Radioactive probes