Learn More
GE Healthcare Amersham™ Hypercassette™ Autoradiography Cassette Available on GSA/VA Contract for Federal Government customers only.
Use for the toughest demands of research in the life sciences. GE Healthcare Life Sciences™ Amersham™ Hypercassette™ Autoradiography Cassettes are constructed from strong aluminium and a choice of colors enables color coding of work.
Manufacturer:  GE Healthcare RPN11648
 View more versions of this product
Catalog No. 45-200-585
Add to cart
Description
Description
Amersham Hypercassette autoradiography cassettes are constructed from strong aluminium and designed for the toughest demands of life science research. Deep cassettes are 1.5cm in depth, allowing them to accommodate thicker samples. A choice of Hypercassette colors enables color coding of work.
  • Provide maximum contact between film and sample
  • Nonporous pad gives a wipe clean surface, ensuring minimum contamination
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Neutral (Standard)
5 x 7 in.
Hypercassette, neutral (standard)
Amersham hypercssette autoradiography cassettes are designed to survive the toughest demands of research in the life sciences.
Light-tight exposures at temperatures below -70°C