GE Healthcare Amersham™ Hypercassette™ Autoradiography Cassette

Use for the toughest demands of research in the life sciences. GE Healthcare Life Sciences™ Amersham™ Hypercassette™ Autoradiography Cassettes are constructed from strong aluminium and a choice of colors enables color coding of work.

Manufacturer: GE Healthcare RPN11644

 View more versions of this product

Catalog No. 45-500-152

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Color Neutral
    Dimensions (L x W) 30 x 40cm
    For Use With (Application) Light-tight exposures at temperatures below -70°C

    Amersham Hypercassette autoradiography cassettes are constructed from strong aluminium and designed for the toughest demands of life science research. Deep cassettes are 1.5cm in depth, allowing them to accommodate thicker samples. A choice of Hypercassette colors enables color coding of work.

    • Provide maximum contact between film and sample
    • Nonporous pad gives a wipe clean surface, ensuring minimum contamination

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit