Learn More
GE Healthcare Amersham™ Hyperscreen™ Intensifying Screen
Offers rapid results and high sensitivity for detecting 32P and 125I in Southern, Northern, and Western blots
Manufacturer:  GE Healthcare RPN1665
 View more versions of this product
Catalog No. 45-000-818
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
13.7 x 16.92 in. (350 x 430mm)
350mm
13.7 in.
16.92 in.
Used with pre-flashed film to give an increase in sensitivity while retaining a linear response that allows quantitation of results
430mm
Autoradiography Screen