Learn More
GE Healthcare Amersham™ Hyperscreen™ Intensifying Screen Available on GSA/VA Contract for Federal Government customers only.
Offers rapid results and high sensitivity for detecting 32P and 125I in Southern, Northern, and Western blots
Manufacturer:  GE Healthcare RPN1669
 View more versions of this product
Catalog No. 45-000-820
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
7.99 in.
10 in.
Used with pre-flashed film to give an increase in sensitivity while retaining a linear response that allows quantitation of results
254mm
Autoradiography Screen
7.99 x 10 in. (203 x 254mm)
203mm