GE Healthcare Amersham™ Megaprime™ DNA Labeling System Available on GSA/VA Contract for Federal Government customers only.
For labeling DNA probes to specific activities >1 x 109dpm/µg in 10 minutes
Manufacturer: GE Healthcare RPN1606
 View more versions of this product
Catalog No. 45-000-809
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With (Application) Any radioactively labeled dCTP
    Quantity 30 Reactions,30 Reactions
    Includes Primer solution 150μL, Labeling Buffer dATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 300μL, Nucleotide solutions dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA, Reaction Buffer A 10 x concentra
    Product Type Labeling System
    Product Name Labeling System
    Kit Contents Primer solution 150μL, Labeling Buffer dATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 300μL, Nucleotide solutions dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA, Reaction Buffer A 10 x concentra
  • Description & Specifications
Related Products
Product Content Feedback