Learn More
GE Healthcare Amersham™ Megaprime™ DNA Labeling System
For labeling DNA probes to specific activities >1 x 109dpm/µg in 10 minutes
Manufacturer:  GE Healthcare RPN1604
 View more versions of this product
Catalog No. 45-000-807
Add to cart
Includes: Primer solution; Labeling buffer; Reaction buffer; Enzyme; Standard DNA solution; Carrier DNA solution
Description
Description
Megaprime™ DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25ng substrate DNA.
  • Features cloned Klenow fragment and random nonamers in the random-prime labeling technique
  • No need to remove unincorporated nucleotides before hybridization
  • Protocols for labeling directly from low melting point agarose
Specifications
Specifications
Membrane hybridization
Megaprime DNA Labeling System, dNTP (for use with any radioactively labeled nucleotide)
-15 - -30°C
Primer solution 150μL; Labeling Buffer dATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 Nucleotide solutions: dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA 120μL, Reaction Buffer A 10 x concentra
30 Reactions