Learn More
GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets Available on GSA/VA Contract for Federal Government customers only.
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Manufacturer:  GE Healthcare 10600075
 View more versions of this product
Catalog No. 45-004-058
  Check Availability
Add to cart
Specifications
Description
Individual sheets.
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Description & Specifications
Specifications
10/Pk.
10cm
Sheets
10cm
Western blots, dot-blot assays, and other protein or nucleic acid methods
0.2um