Learn More
GE Healthcare Amersham Protran NC Nitrocellulose Membranes: Sheets
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. GE Healthcare Amersham Protran NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Manufacturer: GE Healthcare 10600095
Description
Individual sheets.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
25/Pk. | |
0.1 μm | |
9 cm |
Sheet | |
8 cm | |
Western blots, dot-blot assays, and other protein or nucleic acid methods |