Learn More
GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Manufacturer:  GE Healthcare 10600077
 View more versions of this product
Catalog No. 45-005-931
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Individual sheets.
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
Specifications
10/Pk.
0.1 μm
10 cm
Sheet
10 cm
Western blots, dot-blot assays, and other protein or nucleic acid methods