Learn More
GE Healthcare Amersham Protran NC Nitrocellulose Western Blotting Sandwiches
Convenient, precut, preassembled Protran NC Nitrocellulose membrane and filter paper sandwiches
Manufacturer: GE Healthcare 10600104
Description
- Preassembled with two Whatman medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
- Fit most precase gel formats
Specifications
Pack of 10 | |
0.2 μm | |
16 cm |
Sandwich | |
14 cm |