Learn More
GE Healthcare Amersham™ Protran™ NC Nitrocellulose Western Blotting Sandwiches
Convenient, precut, preassembled Protran NC Nitrocellulose membrane and filter paper sandwiches
Manufacturer:  GE Healthcare 10600105
 View more versions of this product
Catalog No. 45-004-082
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
  • Fit most precase gel formats
Specifications
Specifications
Pack of 10
0.1 μm
16 cm
Sandwich
14 cm