GE Healthcare Amersham™ Protran™ NC Nitrocellulose Western Blotting Sandwiches Available on GSA/VA Contract for Federal Government customers only.
Convenient, precut, preassembled Protran NC Nitrocellulose membrane and filter paper sandwiches
Manufacturer: GE Healthcare 10600103
 View more versions of this product
Catalog No. 45-004-080
  Check Availability
Add to Cart
    Specifications
    Format Sandwich, Pk. of 10
    Pore Size 0.45μm
    Length (Metric) 14cm
    Width (Metric) 16cm
  • Description & Specifications
Related Products
Product Content Feedback