GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: GE Healthcare 10600066

Catalog No. 45-004-052

  Check Availability
Add to Cart

    Specifications

    Format Sheets
    Pore Size 0.2μm
    Length (Metric) 16cm
    Width (Metric) 14cm
    For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
    Quantity 25/Pk.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit