Learn More
GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Western Blotting Sandwiches
Precut, preassembled Protran Premium NC Nitrocellulose membrane and filter paper sandwiches save time
Manufacturer:  GE Healthcare 10600107
 View more versions of this product
Catalog No. 45-004-084
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
  • Fit most precase gel formats
Specifications
Specifications
Pack of 10
0.2 μm
16 cm
Sandwich
14 cm