GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Western Blotting Sandwiches Available on GSA/VA Contract for Federal Government customers only.
Precut, preassembled Protran Premium NC Nitrocellulose membrane and filter paper sandwiches save time
Manufacturer: GE Healthcare 10600106
 View more versions of this product
Catalog No. 45-004-083
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Format Sandwich, Pk. of 10
    Pore Size 0.45μm
    Length (Metric) 14cm
    Width (Metric) 16cm
  • Description & Specifications
Related Products
Product Content Feedback