Learn More
GE Healthcare Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets
Provides exceptional handling characteristics, allowing reprobing
Manufacturer:  GE Healthcare 10600036
 View more versions of this product
Catalog No. 45-004-028
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Individual sheets.
  • Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
  • The support does not affect transfer conditions or results
Specifications
Specifications
5/Pk.
Nitrocellulose, Inert Polyester
60 cm
Sheet
0.45 μm
30 cm