GE Healthcare Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets Available on GSA/VA Contract for Federal Government customers only.
Provides exceptional handling characteristics, allowing reprobing
Manufacturer: GE Healthcare 10600068
 View more versions of this product
Catalog No. 45-004-054
  Check Availability
Add to Cart
    Specifications
    Material Nitrocellulose and inert polyester
    Format Sheets
    Pore Size 0.2um
    Length (Metric) 16cm
    Width (English) 14cm
    Quantity 25/Pk.
  • Description & Specifications
Related Products
Product Content Feedback