Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Amersham™ Rapid-hyb Buffer

Pre-mixed, rate-enhancing hybridization buffer. Amersham™ Rapid-hyb Buffer reduces hybridization time for detecting single-copy mammalian genes to just two hours.

Manufacturer:  GE Healthcare RPN1636

 View more versions of this product

Catalog No. 45-000-814


Add to cart

Description

Description

  • For use with radiolabeled DNA, RNA, or oligonucleotide probes
  • Pre-mixed solution improves reproducibility between experiments
  • Suited for Southern and Northern blotting applications
  • Available in two different quantities, sufficient for 1000cm2 membrane and 4000cm2 membrane

Developed for use with nylon membranes; not suitable for use with PVDF or nitrocellulose membranes.

Specifications

Specifications

Radiolabeled DNA, RNA or Oligonucleotide probes
Rapid-hyb Buffer sufficient for 4000 cm2 membrane
500mL
Store at ambient temperature. Under these conditions the product is stable for at least 3 months
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit