Learn More
GE Healthcare Amersham™ Streptavidin-Alkaline phosphatase Conjugate
Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications
Manufacturer:  GE Healthcare RPN1234
Catalog No. 45-000-768
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • High specificity reagents
  • Conjugation achieved using the periodate coupling method described by Nakane and co-workers
  • Sold on the basis of titer in an ELISA and performance in a western blot
Specifications
Specifications
Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry
2mL of a solution containing 10mg/mL BSA, 50mM Tris HCl, 2mM MgCl2 and 0.05% Sodium Azide at pH 8
2mL