GE Healthcare Amersham™ Streptavidin-Fluor Conjugates
Use for in situ hybridization and flow cytometry. GE Healthcare Life Sciences™ Amersham™ Streptavidin-Fluor Conjugates consist of streptavidin conjugated to a range of fluorescent dyes.
Manufacturer: GE Healthcare PA42001
 View more versions of this product
Catalog No. 45-000-728
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Formulation This product is supplied as a lyophilized solid in Phosphate-buffered Saline (0.02M Potassium Phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) Bovine Serum Albumin and 0.1% (w/v) Sodium Azide, reconstitute with 1mL of deionized Water, dispense into suitable aliquots and freeze, avoid repeated freeze-thaw cycles
    For Use With (Application) Suitable for use in the Fluorescent analysis of Biotinylated samples
    Includes Lyophilized solid in phosphate-buffered saline (0.02M potassium phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) bovine serum albumin and 0.1% (w/v) sodium azide, reconstitute with 1mL of deionized water, dispense into suitable aliquots and freeze
    Product Type Cy2-Streptavidin
    Quantity 1mg,1mg
    Product Name Cy2-Streptavidin
    Kit Contents Lyophilized solid in phosphate-buffered saline (0.02M potassium phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) bovine serum albumin and 0.1% (w/v) sodium azide, reconstitute with 1mL of deionized water, dispense into suitable aliquots and freeze
    • High specificity reagents
    Applications:
    in situ hybridization, flow cytometry
  • Description & Specifications
Related Products
Product Content Feedback