Learn More
GE Healthcare Amersham™ TE 77 and TE 77 PWR Semi-Dry Transfer Unit Available on GSA/VA Contract for Federal Government customers only.
Electrotransfer proteins from polyacrylamide gels up to 14 x 16cm (TE 77 PWR) and 21 x 26cm (TE 77) in size in less than one hour
Manufacturer:  GE Healthcare 80621186
 View more versions of this product
Catalog No. 45-001-968
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
The TE 77 and TE 77 PWR offers the convenience of a built-in power supply, and a unique automatic stopping feature. The transfer is stopped when buffer gets depleted, preventing overheating.
  • Long-lasting, perforated platinum-titanium and stainless-steel electrodes give uniform, contamination-free transfers
  • Requires minimal buffer, only enough to saturate blotting papers and membranes
  • Uses low current (0.5A) and voltage (30V) for transfers: safer and more economical
  • Suitable for gels up to 14 x 16cm and 21 x 26cm
  • Tested for use with Amersham™ ECL Western Blotting reagents
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ECL Semi-Dry Blotters are designed to deliver outstanding electrophoretic transfer results using low current and voltage with a minimal amount of buffer. Units are available with and without built-in power supplies to give maximum flexibility.
9 cm
38 cm
21 x 26 cm
25 sheets of blotting paper, 50 sheets of cellophane, solid mask and mask for 14 x 16 cm gels.
TE 77 Semi-Dry Transfer Unit, 21 × 26 cm