GE Healthcare Amersham™ TE 77 and TE 77 PWR Semi-Dry Transfer Unit Available on GSA/VA Contract for Federal Government customers only.
Electrotransfer proteins from polyacrylamide gels up to 14 x 16cm (TE 77 PWR) and 21 x 26cm (TE 77) in size in less than one hour
Manufacturer: GE Healthcare 11001341
 View more versions of this product
Catalog No. 45-001-782
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product.
Please contact customer service for assistance: 1-800-766-7000.
Add to Cart
    Specifications
    Gel Dimensions 14 x 16cm
    Product Type Semi Dry Transfer Unit
    Includes 25 Sheets of blotting Paper, 50 sheets of cellophane, two solid masks and one mask each for 14 x 16cm and 7 x 8cm gels
  • Description & Specifications
Related Products
Product Content Feedback