Learn More
Biochrom™ Tubing for Automatic Sipper
Standard tubing kit includes 6 marprene pump head tubes, PTFE tubing and silicon connection tubing. For use with Ultrospec 7000/8000/9000 sipper.
Manufacturer:  Biochrom™ 80710037
Catalog No. 45-002-324
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Includes:
    • 6 marprene pump head tubes
    • PTFE tubing
    • Silicon connection tubing
  • For use with Ultrospec 7000/8000/9000 sipper
  • Easy to install
Specifications
Specifications
Spectrophotometry
Ultrospec 7000/8000/9000 Spectrophotometer Sipper
Standard tubing
6 marprene pump head tubes, PTFE tubing and silicon connection tubing