Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Biotrak Human High-sensitivity TNFα Assay

Suitable for use with cell culture supernatant, serum and plasma samples

Manufacturer:  GE Healthcare RPN2788

Catalog No. 45-500-158

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Wash buffer, 1.5L

Description

Description

  • Range: 0.3-10pg/mL
  • Sensitivity: 0.1pg/mL
  • Sample volume: 100µL
  • 3.5 hour protocol

Applications:

ELISA

Specifications

Specifications

TNF alpha
0.3 to 10pg/mL
Human High-sensitivity TNFα Assay
0.1pg/mL
Human
ELISA
100μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit