Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Biotrak™ Thromboxane B2 assay

A rapid and sensitive non-isotopic immunoassay method for the determination of thromboxane B2 in plasma and serum samples

Manufacturer:  GE Healthcare RPN220

Catalog No. 45-000-882

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Range: 10-1280pg/mL (0.5-64pg/well)
  • High sensitivity: 0.18pg/well, 3.6pg/mL
  • Two-hour protocol (15-20 hours for maximum sensitivity

Specifications

Specifications

Thromboxane B2
ELISA
Thromboxane B2 assay
1.18 to 3.6pg/mL
2 hr.
10 to 1280pg/mL
Microplate (12 x 8-well strip coated with donkey Anti-Rabbit IgG), Thromboxane B2 peroxidase Conjugate (lyophilized), 2.56ng standard (Thromboxane B2), antiserum (Rabbit Anti-thromboxane B2, lyophilized), TMB Substrate, Assay Buffer concentrate, Wash Buffer concentrate
Each
2° to 8°C
50μL
Safety and Handling

Safety and Handling

ShelfLife : 1 Month

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit