Learn More
GE Healthcare Blue Dextran 2000 Protein Standard
Determine the void fraction in high-resolution gel filtration columns
Manufacturer:  GE Healthcare 17036001
Catalog No. 45-000-048
Add to cart
Description
Description
  • For use in protein isolation and purification
Specifications
Specifications
Blue
Solid
10 g
Determine the void volume in Gel filtration columns
Protein Molecular Weight Standard