Learn More
GE Healthcare Butyl Sepharose™ High Performance Chromatography Media
Designed for intermediate and polishing step purification steps
Manufacturer:  GE Healthcare 17543201
 View more versions of this product
Catalog No. 45-002-954
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
38mg b-lactoglobulin/mL medium
For intermediate and polishing step purifiCatIon steps
Cross-Linked Agarose
25mL
20% Ethanol
24 to 44μm
3 to 13
100 to 200cm/Hour
Butyl
300kPa
Stable in commonly used aqueous Buffers: 1M Sodium Hydroxide, 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Acetonitrile, 70% Ethanol, 3M Ammonium sulfate, 1mM HCl, 30% Isopropanol, 2% SDS
4 to 30°C
High performance Butyl Sepharose media