GE Healthcare Capto™ Multimodal (MMC) Chromatography Media Available on GSA/VA Contract for Federal Government customers only.

Used to solve specific purification challenges at both high and low ionic strengths

Manufacturer: GE Healthcare 17531710

 View more versions of this product

Catalog No. 45-002-953

  Check Availability
Add to Cart

    Specifications

    Anion Exchange Type Weak cation exchanger
    For Use With (Application) For multimodal chromatography
    Matrix Highly cross linked agarose
    Quantity 25mL
    Particle Size 75μm
    Product Type Capto™ MMC media

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit