Learn More
GE Healthcare Cell Proliferation Kit, BioTrak™ ELISA Kit and Components Available on GSA/VA Contract for Federal Government customers only.
Provides a precise, fast and simple colorimetric assay to quantitate proliferating cells cultured in a 96 well microplate
Manufacturer:  GE Healthcare RPN250
 View more versions of this product
Catalog No. 45-000-893
  Check Availability
Add to cart
Specifications
Description
  • Recommended cell number: 10e6-10e7 cells/mL (Cell proliferation kit) 10e2 cells/well-10e5 cells/well (Biotrak ELISA kit)
  • Required time: 2-3 hour post-labeling (labeling 1-2 hours)
  • Antibody and labeling reagent available separately for ELISA and ICC systems
  • Suitable for in vivo and in vitro tissue labeling in addition to cultured cells (ELISA format)
  • Non radioactive protocols provide sensitivity comparable to autoradiographic methods
  • Cell proliferation kit avoids the need to chemically denature DNA. Cellular morphology is preserved allowing multiple labeling experiments
  • 5-fluoro-2'-deoxyuridine is used to inhibit thymidilate synthetase increasing BrdU incorporation
When ordering the Cell Proliferation Kit (RPN20), you must also order DAB REAGENT OF RPN20/RPN1174 (RPNK8888)
Description & Specifications
Specifications
102 to 105 Cells/well
10 x 96 Tests
102 cells/well
Immunocytochemistry
Suspension or Adherent Cells
2° to 8°C