GE Healthcare CyDye™ DIGE Healthcare Fluors: Cy3 Available on GSA/VA Contract for Federal Government customers only.

Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel

Manufacturer: GE Healthcare 25800861

 View more versions of this product

Catalog No. 45-002-252

  Check Availability
Add to Cart

    Specifications

    Product Type Amersham CyDye DIGE Fluor Cy3 Minimal Dye
    Sufficient For For experiments with a limited number of variables, up to 25 labeling
    Storage Requirements -15° to -30°C
    Quantity 2 × 5nM
    Color Green
    Format Solid
    For Use With (Application) For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
    Includes 10nM of CyDye DIGE Fluor Cy3 minimal dye

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit