Learn More
GE Healthcare CyDye™ DIGE Healthcare Fluors: Cy5
Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
Manufacturer:  GE Healthcare 25800862
 View more versions of this product
Catalog No. 45-002-251
Add to cart
Description
Description
  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
  • Allow the labeling of 50µg protein per labeling reaction
For general 2-D applications where sufficient amounts of sample are available.
Specifications
Specifications
Red
For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
For experiments with a limited number of variables, up to 25 labeling
-15° to -30°C
Solid
10nM of CyDye DIGE Fluor Cy5 minimal dye
Amersham CyDye DIGE Fluor Cy5 Minimal Dye
2 × 5nM