Learn More
GE Healthcare DALT Buffer Kit
DALT Buffer Kit for 12 Precast Gels
Manufacturer:  GE Healthcare 17600250
Catalog No. 45-009-999
Add to cart
Description
Description
  • Highest capacity electrophoresis unit with the largest precast gels available (26 u 20 cm)
  • Stable, discontinuous buffer system in a precast, homogeneous gel features high batch-to-batch reproducibility
  • Simple-to-use gel cassettes for gel loading, unloading, and storage
  • Programmable power supply and temperature control ensure high reproducibility among runs.
  • System design minimizes buffer consumption.
Specifications
Specifications
12 Precast Gels