Learn More
GE Healthcare Dithiothreitol (DTT) Available on GSA/VA Contract for Federal Government customers only.
Include reducing agents in the sample solution to break any disulfide bonds present and to maintain all proteins in their fully reduced state. GE Healthcare Life Sciences™ Dithiothreitol (DTT) is one of the most commonly used reductants.
Manufacturer:  GE Healthcare 17131801
 View more versions of this product
Catalog No. 45-000-232
Add to cart
Description
Description
  • Use for IEF, SDS-PAGE
  • White
  • Solid
  • Easily soluble in cold water and hot water
  • Threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
IEF, SDS-PAGE
0 to 5.1
1000mg