GE Healthcare Dithiothreitol (DTT) Available on GSA/VA Contract for Federal Government customers only.

Include reducing agents in the sample solution to break any disulfide bonds present and to maintain all proteins in their fully reduced state. GE Healthcare Life Sciences™ Dithiothreitol (DTT) is one of the most commonly used reductants.

Manufacturer: GE Healthcare 17131802

 View more versions of this product

Catalog No. 45-000-233

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) IEF, SDS-PAGE
    pH 0 to 5.1
    Boiling Point 125° to 130°C
    Quantity 2000mg,2000mg

    • Use for IEF, SDS-PAGE
    • White
    • Solid
    • Easily soluble in cold water and hot water
    • Threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT
     

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit