Learn More
GE Healthcare Ettan DALT Gel, 12.5%
Barcoded for convenient sample and batch tracking. Precast gel cassettes improve handling efficiency and throughput. Gels can be quickly loaded and unloaded with minimal hands-on time
Manufacturer:  GE Healthcare 17600236
Catalog No. 45-009-998
Add to cart
Description
Description
  • Highest capacity electrophoresis unit with the largest precast gels available (26 x 20 cm).
  • Stable, discontinuous buffer system in a precast, homogeneous gel features high batch-to-batch reproducibility.
  • Simple-to-use gel cassettes for gel loading, unloading, and storage.
  • Programmable power supply and temperature control ensure high reproducibility among runs.
  • System design minimizes buffer consumption.
Specifications
Specifications
6/Pk.
Ettan DALTtwelve, Ettan DALTsix Electrophoresis System