Learn More
GE Healthcare Ettan™ DALT Staining Tray Set
For Use With: Ettan DALTtwelve Large Vertical System and Ettan DALTsix Large vertical System
Manufacturer:  GE Healthcare 80646817
Catalog No. 45-005-911
Add to cart
Description
Description
After the second-dimension run is complete, the large-format gels can be stained using DALT staining tray set and PlusOne silver staining kit, Protein. This combination ensures reliable detection with high sensitivity.The set includes two stainless steel trays for staining solutions; one perforated stainless steel tray that seats within the staining trays; and a transparent plastic cover. The perforated insert supports and restrains gels for transfer between staining trays while allowing staining solutions to drain rapidly.
  • For Use With: Ettan DALTtwelve Large Vertical System and Ettan DALTsix Large vertical System
Kit Contains:
  • 2 x Tray for staining solutions (stainless steel)
  • 1 x Perforated tray that seats within the staining trays (23x42 cm, stainless steel)
  • 1 x Cover (transparent plastic)
  • Instructions
Specifications
Specifications
Reproducible high resolution, high throughput 2D electrophoresis
1 Kit
Staining Tray Set
2 Tray for staining solutions, 1 Perforated tray that seats within the staining trays, 1 Plastic cover, instructions
Ettan DALTtwelve Large Vertical System and Ettan DALTsix Large Vertical System