GE Healthcare ExcelGel Precast Gels for SDS-PAGE

Save time with ready-to-use, mechanically stable gels cast on plastic supports. GE Healthcare Life Sciences™ ExcelGel Precast Gels for SDS-PAGE are available as homogeneous or gradient gels.

Manufacturer: GE Healthcare 80126101

 View more versions of this product

Catalog No. 45-001-633

  Check Availability
Add to Cart

    Specifications

    Separation Range 15 kd
    Material Polyacrylamide,Polyester Film
    Length (English) Gel 4.3 in.,4.3 in.
    Buffer Tris Acetic Acid pH 6.4
    Length (Metric) Gel 11cm
    Color Colorless,Colorless
    Stacking Zone 6% T/3% C
    Storage Requirements 2° to 8°C
    Thickness (Metric) 0.05cm
    Width (English) Gel 9.8 in.
    Width (Metric) Gel 25cm,25cm
    For Use With (Application) Analytical chemistry, Research Horizontal electrophoresis of SDS denatured proteins
    Thickness (English) 0.019 in.,0.019 in.
    Format Precast Gel

    Three homogeneous and one gradient polyacrylamide gel for horizontal electrophoresis of SDS denatured proteins; also for separation of nucleic acids in the base pair range 50-1500

    • Homogeneous 7.5, 12.5, and 15% gels have 25 preformed sample wells for sample volumes from up to 17µL for quick, easy sample application
    • Gradient gel uses sample application strips (26 samples up to 40µL, 52 samples up to 20µL), paper pieces, or sample droplets; recommended for use with Immobiline DryStrip gels
    • ExcelGel SDS Homogeneous 15% is also designed for separation of peptides

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit