Learn More
GE Healthcare Factor Xa
Enables the site-specific cleavage of fusion proteins containing an accessible Factor Xa protease recognition sequence such as those expressed in pGEX-X vectors
Manufacturer:  GE Healthcare 27084901
Catalog No. 45-001-321
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Can be used either following affinity purification or while fusion proteins are bound to Glutathione Sepharose™ media
  • Purified from bovine plasma
  • Specifically cleaves at tetrapeptide Ile-Glu-Gly-Arg
Specifications
Specifications
Facto r Xa Protease
400U
Site-specific separation of the GST tag from Proteins expressed using pGEX-X vecto rs, specifically cleaves at Tetrapeptide Ile-Glu-Gly-Arg, one unit cleaves >90% of 100μg of a test GST fusion Protein when incubated in