Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare GammaBind™ G Sepharose™ Medium Available on GSA/VA Contract for Federal Government customers only.

Use recombinant GammaBind™ for laboratory scale antibody purification. GE Healthcare Life Sciences™ GammaBind™ G Sepharose™ Medium binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse

Manufacturer:  GE Healthcare 17088501

 View more versions of this product

Catalog No. 45-000-157


  Check Availability
Add to cart

Specifications

Description

Binds to all IgG subclasses from human, mouse, and rat and total IgG from guinea pig, rabbit, goat, cow, sheep, and horse. Engineered for antibody purification at laboratory scale.

  • Will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin
  • Ligand: GammaBind G type 2 lacking albumin- binding region
  • Ligand coupling method: cyanogen bromide activation
  • Ligand density: » 3mg GammaBind G, type 2/mL medium
  • Number of IgG binding sites per ligand: 2
  • Matrix: Agarose, 4%
  • Average particle size: 90µm
  • Binding capacity: >18 mg human IgG/ml, 6 mg mouse IgG/ml
  • pH stability: 3-9 (working), 2-9 (short term)

Description & Specifications

Specifications

>18mg Human IgG/mL, 6mg Mouse IgG/mL
≈3mg GammaBind G, Type 2/mL drained gel
PBS pH 7, 20% Ethanol
5mL
3-9
For protein purification by affinity coupling
45μm-165μm
0.17 oz.
Sepharose Medium
5mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit