GE Healthcare Gel Casters for MiniVE and SE250, 260 Vertical Units Available on GSA/VA Contract for Federal Government customers only.
For MiniVE, SE250 and SE260 vertical electrophoresis units
Manufacturer: GE Healthcare 80614650
 View more versions of this product
Catalog No. 45-001-711
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) SE 250, SE 260, MiniVE™ vertical electrophoresis system
    Product Type Gel Caster
    Includes 2 Pair of screw clamp assemblies, gaskets and cams
    No. of Gels 1 to 2
  • Description & Specifications
Related Products
Product Content Feedback